Human Complement C1q ELISA Kit
Catalog # ABCAAB170246-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Complement C1q ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Complement C1q ELISA Kit ab170246 is used to quantify Human Complement C1q with a sensitivity of 64 pg/ml. Sandwich ELISA Kit - Validated in a range of biological samples including CSF.