Human IL-10 ELISpot Pair
Katalogové číslo ABCAAB281986-10X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human IL-10 ELISpot pair
- Tests per kit:10×96 tests
Specifications
About this item
Human IL-10 ELISpot pair is a ELISpot pair for the sensitive detection of Human IL-10 production in a single cell suspension.