Human Anti-Dengue virus IgM ELISA Kit
Catalog # ABCAAB108729-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Anti-Dengue virus IgM ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human Anti-Dengue virus IgM ELISA Kit is a Indirect ELISA kit for the measurement of Human Anti-Dengue virus IgM in Human in Citrate plasma, Heparin Plasma, Serum samples.