Order Entry
Czech Republic
ContactUsLinkComponent
Human IL-10 ELISPOT Kit (with un-coated plates)
Human IL-10 ELISPOT Kit (with un-coated plates)
Catalog # ABCAAB46563-480
Supplier:  Abcam
CAS Number:  
undefined
Human IL-10 ELISPOT Kit (with un-coated plates)
Catalog # ABCAAB46563-480
Supplier:  Abcam
CAS Number:  

Specifications

  • Description:
    Human IL-10 ELISPOT kit (with un-coated plates)
  • Seq:
    1
  • Species reactivity:
    Human;Monkey
  • Sample type:
    Suspension cells
  • Form:
    Uncoated
  • Antigen name:
    IL-10
  • Storage temperature:
    +4 °C
  • Processing time:
    Multiple steps
  • Regulatory status:
    RUO

Specifications

About this item

Human IL-10 ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human IL-10 production in a single cell suspension.