Mouse MIP-1 alpha/CCL3 ELISA Kit
Catalog # ABCAAB100726-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:MIP-1 alpha/CCL3
- Description:Mouse MIP-1 alpha/CCL3 ELISA kit
- Sample type:Plasma, cell culture supernatant, serum
- Detection method:Colorimetric
- Detection range:4,1 - 1000 pg/ml
- Storage temperature:4 °C
- Sensitivity:8 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse MIP-1 alpha/CCL3 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse MIP-1 alpha/CCL3 in Mouse in plasma, cell culture supernatant, Serum samples.