Mouse G-CSF Antibody Pair - BSA and Azide free
ABCAAB241682-10X96
: Abcam
:
- Description:Mouse G-CSF Antibody Pair - BSA and Azide free
Mouse G-CSF Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse G-CSF.