Human Complement C4 ELISA Kit
Catalog # ABCAAB108825-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Complement C4 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Human Complement C4 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Complement C4 in Human in Cerebral Spinal Fluid, Milk, Saliva, Urine samples.