Mouse/Rat Retinol Binding Protein 4 Antibody Pair - BSA and Azide Free
ABCAAB241749-10X96
: Abcam
:
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:RBP4
- Description:Mouse/Rat Retinol Binding Protein 4 antibody pair - BSA and azide free
- Detection range:15,62 - 1000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Mouse/Rat Retinol Binding Protein 4 antibody pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Mouse/Rat Retinol Binding Protein 4.