Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Human MMRN1/ECM Antibody Pair - BSA and Azide free
Human MMRN1/ECM Antibody Pair - BSA and Azide free
Human MMRN1/ECM Antibody Pair - BSA and Azide free
Catalog # ABCAAB315402-10X96
Supplier:
Abcam
New Product
CAS Number:
Expand image
Human MMRN1/ECM Antibody Pair - BSA and Azide free
Catalog # ABCAAB315402-10X96
Supplier:
Abcam
New Product
CAS Number:
Specifications
Description:
Human MMRN1/ECM antibody pair - BSA and Azide free
Tests per kit:
10×96 tests
Specifications
About this item
Product Details
Product Details