Human Apolipoprotein AII ELISA Kit (APOA2)
Catalog # ABCAAB108805-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Apolipoprotein AII ELISA Kit (APOA2)
- Tests per kit:96 tests
Specifications
About this item
Human Apolipoprotein AII ELISA Kit (APOA2) is a Sandwich (quantitative) ELISA kit for the measurement of Human Apolipoprotein AII (APOA2) in Human in Plasma, Cell culture extracts, Cell culture supernatant, Serum, Tissue, Cell Lysate samples.