Ovine IL-12 p70 ELISA Kit
Catalog # ABCAAB273255-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Ovine IL-12 p70 ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Ovine IL-12 p70 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Ovine IL-12 p70 in Sheep in cell culture supernatant, serum, EDTA plasma samples.