Order Entry
Czech Republic
Orders LinkContactUsLinkComponent
GFP ELISA Kit

GFP ELISA Kit

New Product
Supplier:   Abcam
GFP ELISA Kit
Supplier:  Abcam
New Product
Certificates
GFP ELISA Kit

3 Options Available Below

Product Details & Documents

GFP ELISA kit is a single-wash 90-min SimpleStep ELISA® used to quantify Green Fluorescence protein with a sensitivity of 2.8 pg/ml. The assay uses a simple Mix-Wash-Read protocol with just one incubation and wash step. Colorimetric assay - 450 nm readout; works on any plate reader. Different formats for different needs: 10×96-well plates for bulk orders and 384-well for higher throughput. Broad sample compatibility, works with tissue lysate and cell lysate.

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • ABCAAB171581-96
  • ABCAAB171581-10X96
  • ABCAAB171581-1X384
  • Target protein:
  • GFP
  • GFP
  • GFP
  • Primary antibody reactivity:
  • Species independent
  • Species independent
  • Species independent
  • Assay Type:
  • Sandwich
  • Sandwich
  • Sandwich
  • Format:
  • Pre-coated
  • Pre-coated
  • Pre-coated
  • Assay duration:
  • One step
  • One step
  • One step
  • Sensitivity:
  • 2,8 pg/ml
  • 2,8 pg/ml
  • 2,8 pg/ml
  • Detection range:
  • 18,8 - 1200 pg/ml
  • 18,8 - 1200 pg/ml
  • 18,8 - 1200 pg/ml
  • Detection method:
  • Colorimetric
  • Colorimetric
  • Colorimetric
  • Sample type:
  • Tissue Lysate, Cell Lysate
  • Tissue Lysate, Cell Lysate
  • Tissue Lysate, Cell Lysate
  • Time to Results:
  • 1 hr 30 min
  • 1 hr 30 min
  • 1 hr 30 min
  • Regulatory status:
  • RUO
  • RUO
  • RUO
  • Storage temperature:
  • 4 °C
  • 4 °C
  • 4 °C

Specifications

Product Family Options

Product Information

  • Tests per kit
    Availability
    Price
  • GFP ELISA Kit
    96 tests
    Log in for availability
    Log in for availability
    1 * 96 Tests
    1 * 96 Tests - 20 090,00 Kč
    20 090,00 Kč
    1 * 96 Tests
       20 090,00 Kč

    Specifications:
    More
    Specifications
    Cat. No.
    ABCAAB171581-96
    Target protein
    GFP
    Primary antibody reactivity
    Species independent
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    One step
    Sensitivity
    2,8 pg/ml
    Detection range
    18,8 - 1200 pg/ml
    Detection method
    Colorimetric
    Sample type
    Tissue Lysate, Cell Lysate
    Time to Results
    1 hr 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
  • GFP ELISA Kit
    10×96 tests
    Log in for availability
    Log in for availability
    1 * 10 Tests
    1 * 10 Tests - 180 600,00 Kč
    180 600,00 Kč
    1 * 10 Tests
       180 600,00 Kč

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCAAB171581-10X96
    Target protein
    GFP
    Primary antibody reactivity
    Species independent
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    One step
    Sensitivity
    2,8 pg/ml
    Detection range
    18,8 - 1200 pg/ml
    Detection method
    Colorimetric
    Sample type
    Tissue Lysate, Cell Lysate
    Time to Results
    1 hr 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    New Product
  • GFP ELISA Kit
    384 tests
    Log in for availability
    Log in for availability
    1 * 1 Tests
    1 * 1 Tests - 50 670,00 Kč
    50 670,00 Kč
    1 * 1 Tests
       50 670,00 Kč

    Specifications:
    New Product
    More
    Specifications
    Cat. No.
    ABCAAB171581-1X384
    Target protein
    GFP
    Primary antibody reactivity
    Species independent
    Assay Type
    Sandwich
    Format
    Pre-coated
    Assay duration
    One step
    Sensitivity
    2,8 pg/ml
    Detection range
    18,8 - 1200 pg/ml
    Detection method
    Colorimetric
    Sample type
    Tissue Lysate, Cell Lysate
    Time to Results
    1 hr 30 min
    Regulatory status
    RUO
    Storage temperature
    4 °C
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.