Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA Kit (HSV1+2)
ABCAAB108742-96
: Abcam
:
- Description:Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA kit (HSV1+2)
- Tests per kit:96 tests
Human Anti-Herpes simplex virus Type 1 and 2 IgM ELISA Kit (HSV1+2) is a Indirect ELISA kit for the measurement of Human Anti-Herpes simplex virus Type 1 and 2 IgM (HSV1+2) in Human in Citrate plasma, Heparin Plasma, Serum samples.