Mouse Prothrombin ELISA Kit
Catalog # ABCAAB157526-96
Supplier: Abcam
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:Prothrombin
- Description:Mouse Prothrombin ELISA kit
- Sample type:Plasma, tissue extracts, cell culture media, serum
- Detection method:Colorimetric
- Detection range:1 - 500 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,386 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifications
About this item
Mouse Prothrombin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Prothrombin in Mouse in plasma, Tissue Extracts, cell culture media, Serum samples.