Human MS2 Antibody Pair - BSA and Azide Free
Catalog # ABCAAB317342-10X96
Supplier: Abcam
New Product
CAS Number:
Specifications
- Assay duration:Multiple steps
- Format:Uncoated
- Target protein:MS2
- Description:Human MS2 antibody pair - BSA and azide free
- Detection range:62,5 - 4000 pg/ml
- Storage temperature:4 °C
- Regulatory status:RUO
- Tests per kit:10×96 tests
Specifications
About this item
Human MS2 antibody pair - BSA and Azide free.