p38 MAPK alpha (pT180/Y182 + Total) ELISA Kit
Catalog # ABCAAB221013-96
Supplier: Abcam
CAS Number:
Specifications
- Description:p38 MAPK alpha (pT180/Y182 + Total) ELISA kit
- Tests per kit:96 tests
Specifications
About this item
p38 MAPK alpha (pT180/Y182 + Total) ELISA kit is a single-wash 90-min Simplestep used to detect p38 MAPK alpha (pT180/Y182 + Total). The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Semi-quantitative sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 5 citations