ContactUsLinkComponent
Mouse CSF-1-R ELISA Kit
Mouse CSF-1-R ELISA Kit
Catalog # ABCAAB213858-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse CSF-1-R ELISA Kit
Catalog # ABCAAB213858-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse CSF-1-R ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse CSF-1-R ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse CSF-1-R in Mouse in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.