ContactUsLinkComponent
SMAD1 (pS463/S465 + Total SMAD1) ELISA Kit
SMAD1 (pS463/S465 + Total SMAD1) ELISA Kit
Catalog # ABCAAB186035-96
Supplier:  Abcam
CAS Number:  
undefined
SMAD1 (pS463/S465 + Total SMAD1) ELISA Kit
Catalog # ABCAAB186035-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    SMAD1 (pS463/S465 + Total SMAD1) ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
SMAD1 (pS463/S465 + Total SMAD1) ELISA Kit is a single-wash 90-min Simplestep used to quantify SMAD1 (pS463/S465 + Total SMAD1) with a sensitivity of 20 µg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.