ContactUsLinkComponent
Mouse GM-CSF ELISA Kit (CSF2)
  ABCAAB100685-96
 :  Abcam
 :  
undefined
Mouse GM-CSF ELISA Kit (CSF2)
  ABCAAB100685-96
 :  Abcam
 :  
  • Description:
    Mouse GM-CSF ELISA Kit (CSF2)
  • Tests per kit:
    96 tests
Mouse GM-CSF ELISA Kit (CSF2) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse GM-CSF (CSF2) in Mouse in Plasma, Cell culture supernatant samples.