ContactUsLinkComponent
Rat TNF alpha ELISA Kit
Rat TNF alpha ELISA Kit
Catalog # ABCAAB108913-96
Supplier:  Abcam
CAS Number:  
undefined
Rat TNF alpha ELISA Kit
Catalog # ABCAAB108913-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Rat TNF alpha ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Rat TNF alpha ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Rat TNF alpha in Rat in cell culture supernatant samples.