ContactUsLinkComponent
Pig Plasminogen ELISA Kit
Pig Plasminogen ELISA Kit
Catalog # ABCAAB190538-96
Supplier:  Abcam
CAS Number:  
undefined
Pig Plasminogen ELISA Kit
Catalog # ABCAAB190538-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Pig Plasminogen ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Pig Plasminogen ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Pig Plasminogen in Pig in Plasma, Serum samples.