ContactUsLinkComponent
Mouse IL-12 p70 ELISA Kit
Mouse IL-12 p70 ELISA Kit
Catalog # ABCAAB119531-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse IL-12 p70 ELISA Kit
Catalog # ABCAAB119531-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse IL-12 p70 ELISA kit
Specifications
About this item
Mouse IL-12 p70 ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse IL-12 p70 in Mouse in cell culture supernatant, serum samples.