Human Stanniocalcin 2 / STC-2 ELISA Kit
Catalog # ABCAAB222880-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Stanniocalcin 2 / STC-2 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Human Stanniocalcin 2 / STC-2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human Stanniocalcin 2 / STC-2 in Human in Plasma, Serum samples.