Mouse NBL1 ELISA Kit
Catalog # ABCAAB234575-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse NBL1 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Mouse NBL1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse NBL1 with a sensitivity of 21,6 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.