Doxycycline ELISA Kit
ABCAAB285232-96
: Abcam
:
- Pk:96 Tests
- Assay duration:Multiple steps
- Assay Type:Competitive
- Format:Pre-coated
- Target protein:Doxycycline
- Description:Doxycycline ELISA kit
- Detection method:Colorimetric
- Detection range:0,1 - 8,1 ng/ml
- Storage temperature:4 °C
- Sensitivity:0,1 ng/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Doxycycline ELISA kit is a Competitive ELISA kit for the measurement of Doxycycline in Tissue, Milk, Serum samples.