Mouse MUP1 ELISA Kit
Catalog # ABCAAB284028-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse MUP1 ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse MUP1 ELISA kit is a sandwich (quantitative) ELISA kit for the measurement of Mouse MUP1 in Mouse in Urine, plasma, serum samples.