ContactUsLinkComponent
Human Complement C5a ELISA Kit
Human Complement C5a ELISA Kit
Catalog # ABCAAB193695-96
Supplier:  Abcam
CAS Number:  
undefined
Human Complement C5a ELISA Kit
Catalog # ABCAAB193695-96
Supplier:  Abcam
CAS Number:  
About this item
Human Complement C5a ELISA kit ab193695 is used to quantify Human Complement C5a with a sensitivity of 0,17 ng/ml. Sandwich ELISA kit.