Mouse BAFF ELISA Kit
ABCAAB216948-96
: Abcam
:
- Description:Mouse BAFF ELISA kit
Mouse BAFF ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse BAFF with a sensitivity of 7.4 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.