Human Interferon gamma + IL-10 ELISPOT Set
Catalog # ABCAAB48722-1920
Supplier: Abcam
CAS Number:
Specifications
- Description:Human Interferon gamma + IL-10 ELISPOT set
- Species reactivity:Human
- Sample type:Suspension cells
- Form:Uncoated
- Storage temperature:+4 °C
- Processing time:Multiple steps
- Antigen symbol:Interferon gamma + IL10
- Regulatory status:RUO
Specifications
About this item
Human Interferon gamma + IL-10 ELISPOT Set is a ELISPOT Set for the sensitive detection of Human Interferon gamma + IL-10 production in a single cell suspension.