ContactUsLinkComponent
Human CD62P ELISA Kit (P-Selectin)
Human CD62P ELISA Kit (P-Selectin)
  ABCAAB100631-96
 :  Abcam
 :  
undefined
Human CD62P ELISA Kit (P-Selectin)
  ABCAAB100631-96
 :  Abcam
 :  
  • Description:
    Human CD62P ELISA kit (P-Selectin)
Human CD62P ELISA kit (P-Selectin) is a Sandwich (quantitative) ELISA kit for the measurement of Human CD62P (P-Selectin) in Human in plasma, cell culture supernatant, serum samples.