Human alpha 2 Macroglobulin ELISA Kit
Catalog # ABCAAB108883-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Human alpha 2 Macroglobulin ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Human alpha 2 Macroglobulin ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human alpha 2 Macroglobulin in Human in cerebral spinal fluid, saliva cell culture supernatant, milk samples.