Human MS2 ELISA Kit
ABCAAB213750-96
: Abcam
:
- Description:Human MS2 ELISA kit
- Tests per kit:96 tests
Human MS2 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human MS2 in Human in Cell culture supernatant, Serum samples.