Rat BNP 32 ELISA Kit
Catalog # ABCAAB108815-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Rat BNP 32 ELISA Kit
- Tests per kit:96 tests
Specifications
About this item
Rat BNP 32 ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat BNP 32 in Rat in Cell culture supernatant, Plasma, Serum, Tissue samples.