Mouse Troponin I ELISA Kit
Catalog # ABCAAB285235-96
Supplier: Abcam
CAS Number:
Specifications
- Pk:96 Tests
- Assay Type:Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Troponin I ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Troponin I ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Troponin I in Mouse in Plasma, Serum, Other biological fluids samples.