Order Entry
Czech Republic
ContactUsLinkComponent
Rat Activin A ELISA Kit
Rat Activin A ELISA Kit
Catalog # ABCAAB119570-96
Supplier:  Abcam
CAS Number:  
undefined
Rat Activin A ELISA Kit
Catalog # ABCAAB119570-96
Supplier:  Abcam
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Target protein:
    Activin A
  • Description:
    Rat Activin A ELISA kit
  • Sample type:
    Tissue extracts, cell culture supernatant, serum, other biological fluids
  • Detection method:
    Colorimetric
  • Detection range:
    62,5 - 4000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    12 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

Specifications

About this item

Rat Activin A ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Rat Activin A in Rat in Tissue Extracts, cell culture supernatant, Serum, Other biological fluids samples.