ContactUsLinkComponent
Human IL-1 beta ELISPOT Kit (with un-coated plates)
Human IL-1 beta ELISPOT Kit (with un-coated plates)
  ABCAAB46608-480
 :  Abcam
 :  
undefined
Human IL-1 beta ELISPOT Kit (with un-coated plates)
  ABCAAB46608-480
 :  Abcam
 :  
  • Description:
    Human IL-1 beta ELISPOT kit (with un-coated plates)
  • Tests per kit:
    5×96 tests
Human IL-1 beta ELISPOT Kit (with un-coated plates) is a ELISPOT kit for the sensitive detection of Human IL-1 beta production in a single cell suspension.