Human SDF1 beta ELISA Kit
Katalogové číslo ABCAAB314389-96TST
Dodavatel: Abcam
Číslo CAS:
Specifikace
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:SDF1 beta
- Description:Human SDF1 beta ELISA kit
- Detection method:Colorimetric
- Detection range:35 - 8000 pg/ml
- Storage temperature:4 °C
- Sensitivity:35 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Specifikace
O tomto produktu
Human SDF1 beta ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Human SDF1 beta in Human in Serum, plasma, cell culture supernatant samples.