Mouse Flt 3 Ligand ELISA Kit
Catalog # ABCAAB113342-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse Flt 3 Ligand ELISA kit
- Tests per kit:96 tests
Specifications
About this item
Mouse Flt 3 Ligand ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse Flt 3 Ligand in Mouse in Plasma, Cell culture supernatant, Serum samples.