Mouse G-CSF ELISA Kit (CSF3)
Catalog # ABCAAB100684-96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse G-CSF ELISA kit (CSF3)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse G-CSF ELISA kit (CSF3) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse G-CSF (CSF3) in Mouse in plasma, cell culture supernatant, serum samples.