Human DKK3 ELISA Kit (Dickkopf-3)
ABCAAB100502-96
: Abcam
:
- Description:Human DKK3 ELISA Kit (Dickkopf-3)
- Tests per kit:96 tests
Human DKK3 ELISA Kit (Dickkopf-3) is a Sandwich (quantitative) ELISA kit for the measurement of Human DKK3 (Dickkopf-3) in Human in Plasma, Cell culture supernatant, Serum samples.