ContactUsLinkComponent
Mouse C Reactive Protein ELISA Kit
Mouse C Reactive Protein ELISA Kit
Catalog # ABCAAB222511-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse C Reactive Protein ELISA Kit
Catalog # ABCAAB222511-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse C Reactive Protein ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse C Reactive Protein ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse C Reactive Protein with a sensitivity of 17 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.