Rat Fractalkine ELISA Kit
ABCAAB229890-1X96
: Abcam
:
- Description:Rat Fractalkine ELISA kit
- Tests per kit:96 tests
Rat Fractalkine ELISA Kit is a single-wash 90-min Simplestep used to quantify Rat Fractalkine with a sensitivity of 9,8 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.