ContactUsLinkComponent
p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha ELISA Kit
p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha ELISA Kit
Catalog # ABCAAB126453-96
Supplier:  Abcam
CAS Number:  
undefined
p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha ELISA Kit
Catalog # ABCAAB126453-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha ELISA Kit is a Semi-quantitative ELISA kit for the measurement of p38 MAPK alpha (pT180/pY182) + total p38 MAPK alpha in Mouse, Human in Cell Lysate samples.