Mouse SLPI ELISA kit (ALP)
Catalog # ABCAAB267583-1X96
Supplier: Abcam
CAS Number:
Specifications
- Description:Mouse SLPI ELISA kit (ALP)
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
Mouse SLPI ELISA kit (ALP) is a Sandwich (quantitative) ELISA kit for the measurement of Mouse SLPI (ALP) in Mouse in plasma, cell culture supernatant, serum samples.