ContactUsLinkComponent
11-Dehydro-TXB2 ELISA Kit
11-Dehydro-TXB2 ELISA Kit
Catalog # ABCAAB133054-96
Supplier:  Abcam
CAS Number:  
undefined
11-Dehydro-TXB2 ELISA Kit
Catalog # ABCAAB133054-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    11-Dehydro-TXB2 ELISA Kit
  • Tests per kit:
    96 tests
Specifications
About this item
11-dehydro-TXB2 ELISA Kit is a Competitive ELISA kit for the measurement of 11-dehydro-TXB2 in Human in Urine, Cell culture supernatant samples.