ContactUsLinkComponent
Acetyl Coenzyme A carboxylase alpha ELISA Kit
Acetyl Coenzyme A carboxylase alpha ELISA Kit
Catalog # ABCAAB309323-96TST
Supplier:  Abcam
CAS Number:  
undefined
Acetyl Coenzyme A carboxylase alpha ELISA Kit
Catalog # ABCAAB309323-96TST
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Acetyl Coenzyme A carboxylase alpha ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Acetyl Coenzyme A carboxylase alpha ELISA kit is a single-wash 90-min Simplestep used to quantify Acetyl Coenzyme A carboxylase alpha with a sensitivity of 25,132 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric sandwich ELISA - 450 nm readout : works on any standard plate reader - Design your own immunoassay: we also offer the conjugation-ready antibody pair.