Mouse TNF alpha ELISA Kit
ABCAAB100747-96
: Abcam
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Target protein:TNF alpha
- Description:Mouse TNF alpha ELISA kit
- Sample type:Serum free media, plasma, cell culture supernatant
- Detection method:Colorimetric
- Detection range:93,75 - 6000 pg/ml
- Storage temperature:4 °C
- Sensitivity:60 pg/ml
- Regulatory status:RUO
- Tests per kit:96 tests
Mouse TNF alpha ELISA kit is a Sandwich (quantitative) ELISA kit for the measurement of Mouse TNF alpha in Mouse in serum free media, plasma, cell culture supernatant samples.