Mouse MCP1 ELISA Kit
Supplier: Abcam
Certificates
About this item
Mouse MCP1 ELISA kit is a single-wash 90-min Simplestep used to quantify Mouse MCP1 with a sensitivity of 0,487 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader. Different formats for different needs: 10×96 plates for bulk orders and 384-well plate for higher throughput.
Specifications
- Storage temperature:+4 ℃