HLA-E ELISA Kit
Catalog # ABCAAB300337-96
Supplier: Abcam
CAS Number:
Specifications
- Description:HLA-E ELISA kit
- Storage temperature:+4 ℃
- Tests per kit:96 tests
Specifications
About this item
HLA-E ELISA kit is a single-wash 90-min Simplestep used to quantify HLA-E with a sensitivity of 25,806 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. Colorimetric Sandwich ELISA - 450 nm readout: works on any standard plate reader.