ContactUsLinkComponent
Mouse YKL-40 / CHI3L1 ELISA Kit
Mouse YKL-40 / CHI3L1 ELISA Kit
Catalog # ABCAAB238262-96
Supplier:  Abcam
CAS Number:  
undefined
Mouse YKL-40 / CHI3L1 ELISA Kit
Catalog # ABCAAB238262-96
Supplier:  Abcam
CAS Number:  
Specifications
  • Description:
    Mouse YKL-40 / CHI3L1 ELISA kit
  • Tests per kit:
    96 tests
Specifications
About this item
Mouse YKL-40 / CHI3L1 ELISA Kit is a single-wash 90-min Simplestep used to quantify Mouse YKL-40 / CHI3L1 with a sensitivity of 3,17 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader.